Learn More
Cayman Chemical GLP-21-34humntrIfluo 1mg
SDP
Catalog No. 503015686
Encompass_Preferred
A synthetic form of the peptide GLP-2; increases survival of myenteric neurons cultured alone (EC50 = 0.35 nM) and when co-cultured with mast cells to induce cell death
Catalog No. 50-301-5686 Supplier Cayman Chemical Supplier No. 247561 MG
A synthetic form of the peptide GLP-2; increases survival of myenteric neurons cultured alone (EC50 = 0.35 nM) and when co-cultured with mast cells to induce cell death