Learn More
Medchemexpress LLC HY-124614 5mg Medchemexpress, GLP-26 CAS:2133017-36-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1246145MG
Encompass_Preferred
Medchemexpress, HY-124614 5mg GLP-26 CAS:2133017-36-2 GLP-26 is a HBV capsid assembly modulators (CAM), inhibits HBV DNA replication in Hep AD38 system (IC50=3 nM), and reduces cccDNA by >90% at 1 μM.GLP-26 disrupts the encapsidation of pre-genomic RNA, causes nucleocapsid disassembly and reduces cccDNA pools. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9669