Learn More
Cayman Chemical GLP-2ratrIfluoroacetate 1mg
SDP
Catalog No. 502936217
Encompass_Preferred
A gut peptide hormone agonist of GLP-2R; increases cAMP levels in COS cells expressing GLP-2R (EC50 = 0.58 nM); produced by enteroendocrine L cells of the small and large intestine and in the hypothalamus; increases small bowel weight by 50% and increases villus height and crypt cell proliferation in the small intestine of mice (43.75 μg twice per day for 10 days); reduces two-hour food intake in rats by 37% (10 μg); secretion is stimulated by dietary carbohydrates, glucose, dextrin, and oils rich in polyunsaturated or monounsaturated fatty acids
Catalog No. 50-293-6217 Supplier Cayman Chemical Supplier No. 247571 MG
A gut peptide hormone agonist of GLP-2R; increases cAMP levels in COS cells expressing GLP-2R (EC50 = 0.58 nM); produced by enteroendocrine L cells of the small and large intestine and in the hypothalamus; increases small bowel weight by 50% and increases villus height and crypt cell proliferation in the small intestine of mice (43.75 μg twice per day for 10 days); reduces two-hour food intake in rats by 37% (10 μg); secretion is stimulated by dietary carbohydrates, glucose, dextrin, and oils rich in polyunsaturated or monounsaturated fatty acids