Learn More
Medchemexpress LLC Glp-Asn-Pro-AMC | 291752-43-7 | 99.3% | 497.50 | 25 MG
SDP
Catalog No. 5000423864
Encompass_Preferred
Glp-Asn-Pro-AMC is identified as an inhibitor of thyrotropin-releasing hormone degrading enzyme (TRH-DE), exhibiting a Ki value of 0.97 μM.
  • Inhibitor of thyrotropin-releasing hormone degrading enzyme (TRH-DE)
Catalog No. 50-004-23864 Supplier Medchemexpress LLC Supplier No. HYP10485100MG
Glp-Asn-Pro-AMC is identified as an inhibitor of thyrotropin-releasing hormone degrading enzyme (TRH-DE), exhibiting a Ki value of 0.97 μM.
  • Inhibitor of thyrotropin-releasing hormone degrading enzyme (TRH-DE)