Learn More
Cayman Chemical Glp1-ApelIn-13trIfluoroac 1mg
SDP
Catalog No. 502970847
Encompass_Preferred
A pyroglutamylated form of apelin-13, in that the N-terminal glutamine residue is cyclized to pyroglutamic acid; a potent APJ receptor agonist (EC50 = 0.30 nM); the major apelin isoform in the plasma of healthy human subjects
Catalog No. 50-297-0847 Supplier Cayman Chemical Supplier No. 155901 MG
A pyroglutamylated form of apelin-13, in that the N-terminal glutamine residue is cyclized to pyroglutamic acid; a potent APJ receptor agonist (EC50 = 0.30 nM); the major apelin isoform in the plasma of healthy human subjects