Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GLP1R protein, human (HEK293, N-His, C-Myc) | >95.0% | 100UG
SDP

Catalog No. 5000175448
Encompass_Preferred

Recombinant human GLP1R protein expressed in HEK293 cells and supplied lyophilized with N-terminal His and C-terminal C-Myc tags. The glycosylated receptor is functional in ligand-binding assays, activates adenylyl cyclase to increase intracellular cAMP upon ligand binding, and is suitable for biochemical characterization, antibody validation, and receptor-ligand interaction studies.

  • Expressed in HEK293 cells.
  • N-His and C-Myc tagged for detection and purification.
  • Glycosylated; apparent molecular weight ~25-38 kDa.
  • Purity >95% by reducing SDS-PAGE.
  • Available in 20 μg, 50 μg, and 100 μg quantities.
  • Lyophilized formulation with trehalose for stability.
  • Store at -20°C; reconstituted material stable at 4°C for one week.

Catalog No. 50-001-75448 Supplier Medchemexpress LLC Supplier No. HYP700468100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human GLP1R protein expressed in HEK293 cells and supplied lyophilized with N-terminal His and C-terminal C-Myc tags. The glycosylated receptor is functional in ligand-binding assays, activates adenylyl cyclase to increase intracellular cAMP upon ligand binding, and is suitable for biochemical characterization, antibody validation, and receptor-ligand interaction studies.

  • Expressed in HEK293 cells.
  • N-His and C-Myc tagged for detection and purification.
  • Glycosylated; apparent molecular weight ~25-38 kDa.
  • Purity >95% by reducing SDS-PAGE.
  • Available in 20 μg, 50 μg, and 100 μg quantities.
  • Lyophilized formulation with trehalose for stability.
  • Store at -20°C; reconstituted material stable at 4°C for one week.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.