Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glucagon-like peptide-1 receptor, human (HEK293), C-terminal Fc fusion | >95.0% | 5UG
SDP

Supplier:  Medchemexpress LLC HYP703525UG

Encompass_Preferred

Recombinant human glucagon-like peptide-1 receptor (GLP-1R) expressed in HEK293 cells and provided as a C-terminal human Fc fusion. Supplied as a lyophilized powder with high purity and low endotoxin for receptor binding, signaling studies, assay development, and antibody characterization. Intended for research use only.

  • Recombinant human GLP-1 receptor expressed in HEK293 cells.
  • C-terminal human Fc fusion for improved stability and detection.
  • Lyophilized powder for long-term storage and transport.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg suitable for many in vitro assays.
  • Do not reconstitute below 100 μg/mL; freeze aliquots for extended storage.

Catalog No. 50-001-76266


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.