Learn More
Eurofins Discoverx PathHunter GLP1R Total GPCR Internalization U2OS Cell Line, 2 vials. (93-1075C3)
Catalog No. 502001985
Encompass
The PathHunter GLP1R Total GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of GLP1R via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged GLP1R and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the GLP1R receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1985 Supplier Eurofins Discoverx Supplier No. 931075C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter GLP1R Total GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of GLP1R via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged GLP1R and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the GLP1R receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.