Learn More
Eurofins Discoverx PathHunter GLP2R Bioassay Kit, 2-Plate. (93-0572Y2-00133)
Catalog No. 502000867
Encompass
The PathHunter GLP2R Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. GLP2) based activation of GLP2R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GLP2R and EA-tagged β-Arrestin-2. Activation of GLP2R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-0867 Supplier Eurofins Discoverx Supplier No. 930572Y200133
The PathHunter GLP2R Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. GLP2) based activation of GLP2R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GLP2R and EA-tagged β-Arrestin-2. Activation of GLP2R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.