Learn More
Quip Laboratories Inc GLPC Acid, 5 Gallon
SDP
Catalog No. NC9070812
Encompass
GLPC-Acid is a liquid blend of organic acids perfect for decontaminating scientific glassware. This product also solves a wide range of soil removal challenges on other surfaces.
This item is not returnable. View return policy
Catalog No. NC9070812 Supplier Quip Laboratories Inc Supplier No. GLPAC5GAL
GLPC-Acid is a liquid blend of organic acids perfect for decontaminating scientific glassware. This product also solves a wide range of soil removal challenges on other surfaces.