Learn More
TARGETMOL CHEMICALS INC GLPG0492 5MG
SDP
Catalog No. 502581532
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GLPG0492 is a novel selective androgen receptor modulator. After oral dosing it showed anabolic activity on muscle strongly dissociated from the androgenic activity on the prostate. purity: 98%
Catalog No. 50-258-1532 Supplier TARGETMOL CHEMICALS INC Supplier No. T11410L5MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GLPG0492 is a novel selective androgen receptor modulator. After oral dosing it showed anabolic activity on muscle strongly dissociated from the androgenic activity on the prostate. purity: 98%