Learn More
Medchemexpress LLC HY-18102 5mg Medchemexpress, GLPG0492 CAS:1215085-92-9 Purity:>98%
SDP
Catalog No. 502082018
Encompass_Preferred
Medchemexpress, HY-18102 5mg GLPG0492 CAS:1215085-92-9 GLPG0492 is a non-steroidal selective androgen receptor modulator (potency 12 nM). GLPG0492 has the potential for the research of musculo-skeletal diseases such as sarcopenia and cachexia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2018 Supplier Medchemexpress LLC Supplier No. HY181025MG
Medchemexpress, HY-18102 5mg GLPG0492 CAS:1215085-92-9 GLPG0492 is a non-steroidal selective androgen receptor modulator (potency 12 nM). GLPG0492 has the potential for the research of musculo-skeletal diseases such as sarcopenia and cachexia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.