Learn More
TARGETMOL CHEMICALS INC GLPG0634 ANALOG 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T307610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GLPG0634 analog (GLPG0634 analogue) is a specific JAK1 inhibitor with IC50 of 10/28/810/116 nM for JAK1/2/3 and TYK2 respectively. purity: 99%
Catalog No. 50-258-0392