Learn More
Selleck Chemical LLC GLPG1837 2mg 1654725-02-6 ABBV-974
SDP
Supplier:  Selleck Chemical LLC S86982MG
Encompass_Preferred
GLPG1837 (ABBV-974) is a novel CFTR potentiator with an EC50 value of 3 nM for F508del, showing enhanced efficacy on CFTR mutants harboring class III mutations compared to Ivacaftor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8226