Learn More
Medchemexpress LLC HY-119936 5mg Medchemexpress, GLPG2451 CAS:2055015-61-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1199365MG
Encompass_Preferred
Medchemexpress, HY-119936 5mg GLPG2451 CAS:2055015-61-5 GLPG2451 is a cystic fibrosis transmembrane conductance regulator (CFTR) potentiator, which effectively potentiates low temperature rescued F508del CFTR with an EC50 of 11.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9556