Learn More
Medchemexpress LLC GLPG3970 | 2403733-82-2 | C25H27F3N4O4 | 250 MG
SDP
Catalog No. 5000375956
Encompass_Preferred
GLPG3970 is a first-in-class SIK2/SIK3 inhibitor intended for research related to inflammation and autoimmune diseases. It has been investigated in clinical trials for conditions such as Systemic Lupus Erythematosus, Ulcerative Colitis, and Rheumatoid Arthritis.
  • Acts as a SIK2/SIK3 inhibitor
  • Supports research in inflammation and autoimmune diseases
  • Induces IL-10 expression in human CD14+ monocytes with an EC50 of 1087 nM
  • Reduces LPS-stimulated TNF-alpha production in human CD14+ monocytes with an IC50 of 133 nM
  • Decreases ear thickness in murine models of psoriatic-like epidermal hyperplasia
  • Improves clinical manifestations in murine collagen-antibody induced arthritis models
  • Reduces disease activity index in mice T cell transfer models
  • Investigated in clinical trials for various conditions
  • High purity: 99.70%
  • Solid, white to off-white appearance
Catalog No. 50-003-75956 Supplier Medchemexpress LLC Supplier No. 250MGHY150203
GLPG3970 is a first-in-class SIK2/SIK3 inhibitor intended for research related to inflammation and autoimmune diseases. It has been investigated in clinical trials for conditions such as Systemic Lupus Erythematosus, Ulcerative Colitis, and Rheumatoid Arthritis.
  • Acts as a SIK2/SIK3 inhibitor
  • Supports research in inflammation and autoimmune diseases
  • Induces IL-10 expression in human CD14+ monocytes with an EC50 of 1087 nM
  • Reduces LPS-stimulated TNF-alpha production in human CD14+ monocytes with an IC50 of 133 nM
  • Decreases ear thickness in murine models of psoriatic-like epidermal hyperplasia
  • Improves clinical manifestations in murine collagen-antibody induced arthritis models
  • Reduces disease activity index in mice T cell transfer models
  • Investigated in clinical trials for various conditions
  • High purity: 99.70%
  • Solid, white to off-white appearance