Learn More
ABclonal Technology GLRA2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A156731000UL
Encompass_Preferred
The glycine receptor consists of two subunits, alpha and beta, and acts as a pentamer. The protein encoded by this gene is an alpha subunit and can bind strychnine. Several transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-05298