Learn More
ABclonal Technology GLRA2 Rabbit pAb
SDP
Catalog No. 5000107947
Encompass_Preferred
The glycine receptor consists of two subunits, alpha and beta, and acts as a pentamer. The protein encoded by this gene is an alpha subunit and can bind strychnine. Several transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-07947 Supplier ABclonal Technology Supplier No. A15673500UL
The glycine receptor consists of two subunits, alpha and beta, and acts as a pentamer. The protein encoded by this gene is an alpha subunit and can bind strychnine. Several transcript variants encoding different isoforms have been found for this gene.