Learn More
Aviva Systems Biology Corp Glrx1 Antibody - middle region (ARP54625_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54625_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Glrx1. It was validated on Western Blot. . Alternate names: Glrx, Grx, Glrx1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-285-948