Learn More
Aviva Systems Biology Corp GLRX2 Antibody - middle region (ARP57792_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57792_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLRX2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GLRX2. Alternate names: GRX2, bA101E13.1, CGI-133ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-286-379