Learn More
Cayman Chemical GLS1 InhIbItr 1mg
SDP
Catalog No. 502939806
Encompass_Preferred
An inhibitor of GLS1 (IC50 = 0.021 μM); inhibits the growth of NCI H1703 NSCLC cells in vitro (GI50 = 0.011 μM); reduces tumor growth, increases tumor levels of glutamine, and decreases tumor levels of glutamate and aspartate in an NCI H1703 mouse xenograft model at 100 mg/kg
Catalog No. 50-293-9806 Supplier Cayman Chemical Supplier No. 285811 MG
Add to Cart
Add to Cart
An inhibitor of GLS1 (IC50 = 0.021 μM); inhibits the growth of NCI H1703 NSCLC cells in vitro (GI50 = 0.011 μM); reduces tumor growth, increases tumor levels of glutamine, and decreases tumor levels of glutamate and aspartate in an NCI H1703 mouse xenograft model at 100 mg/kg