Learn More
Carbosynth LLC. Glt-Ala-Ala-Pro-Leu-pNA H2O, 0.1g, MW: 604.65 18.02, Molecular formula: C28H40N6O9 H2O
SDP
Supplier:  Carbosynth LLC. SGL31290.1G
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Glutaryl-L-Alanyl-L-Alanyl-L-Prolyl-L-Leucine-p-Nitroanilide, Purity: , Grade AA, Application: Substrate for Pancreatic Elastase, Storage Temperature: -20C, References: E.G. Del Mar, C. Largman, J.W. Brodrick, M. Fassett, and M.C. Geokas, Biochemistry, 19, 468 (1980).
Catalog No. 50-168-7107