Learn More
Aviva Systems Biology Corp GLT8D1 Antibody - middle region (ARP44608_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44608_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLT8D1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GLT8D1. Alternate names: AD-017, DKFZp781O20198, FLJ14611, MSTP139ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58859