Learn More
Aviva Systems Biology Corp GLT8D2 Antibody - C-terminal region (ARP49993_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49993_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLT8D2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GLT8D2. Alternate names: FLJ31494ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53613