Learn More
Cayman Chemical γ-Glu-Ala (trifluoroacetate salt), Purity: ≥95%, 250 mg
SDP
Catalog No. 502913791
Encompass_Preferred
A dipeptide; has been found in P. roqueforti; has been found in ripened Gouda, goat, and Milner cheeses; a positive allosteric modulator of CaSR (EC50 = 4.8 µM in CaSR-expressing HEK293 cells); induces glutathionuria in mice at 4 mmol/kg; plasma levels are increased in malnourished neonatal mice; increased serum levels are positively associated with an increased risk of chronic kidney disease
Catalog No. 50-291-3791 Supplier Cayman Chemical Supplier No. 36688250 MG
A dipeptide; has been found in P. roqueforti; has been found in ripened Gouda, goat, and Milner cheeses; a positive allosteric modulator of CaSR (EC50 = 4.8 µM in CaSR-expressing HEK293 cells); induces glutathionuria in mice at 4 mmol/kg; plasma levels are increased in malnourished neonatal mice; increased serum levels are positively associated with an increased risk of chronic kidney disease