Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals [Glu1] Fibrinopeptide B, Human

Catalog No. AAJ66755MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66755MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66755MCR Supplier Thermo Scientific Chemicals Supplier No. J66755MCR
Add to Cart

1570.57 g/mol

[Glu^1] Fibrinopeptide B is used as a indicator of coagulation and fibrinolytic activity. And is useful for the tuning and calibration of electrospray ionization and MALDI mass spectrometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Species Human
Purification Method HPLC
Common Name [Glu1] Fibrinopeptide B
Content And Storage -30°C to -10°C
Format Powder
Molecular Weight (g/mol) 1570.57
Product Type [Glu1] Fibrinopeptide B, human
Quantity 1 mg
Regulatory Status RUO
Sequence Glu-Gly-Val-Asn-Asp-Asn-Glu-Glu-Gly-Phe-Phe-Ser-Ala-Arg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.