Learn More
R&D SYSTEMS HEMATOLOGY HMQHGB201, HEGLU
Catalog No. NC0252656
Encompass_Preferred
The HMQHGB201 is a digital handheld glucose meter from RnDHeme that measures blood glucose levels. It has a measurement range of 20-600 mg/dL and uses an electrochemical test strip method. This meter requires only a small 0.5 μL blood sample size. It has a test time of 5 seconds and memory capacity for 250 test results. The HMQHGB201 includes 10 free test strips, 10 free lancets, a lancing device, and a carrying case. This meter is CE and ISO certified.
This item is not returnable. View return policy
Catalog No. NC0252656 Supplier R&D SYSTEMS HEMATOLOGY Supplier No. GH00HX
The HMQHGB201 is a digital handheld glucose meter from RnDHeme that measures blood glucose levels. It has a measurement range of 20-600 mg/dL and uses an electrochemical test strip method. This meter requires only a small 0.5 μL blood sample size. It has a test time of 5 seconds and memory capacity for 250 test results. The HMQHGB201 includes 10 free test strips, 10 free lancets, a lancing device, and a carrying case. This meter is CE and ISO certified.