Learn More
Cayman Chemical -Glu-PhetrIfluoroACTAte100mg
SDP
Catalog No. 502911605
Encompass_Preferred
A dipeptide with metabolism-altering activity; prevents increases in lipid accumulation in oleic acid-stimulated AML12 hepatocytes at 30 µM; increases CaSR levels and secretion of CCK and GLP-1 in STC-1 enteroendocrine cells at 2.5-10 mM; changes in urine levels are associated with hypertension and PKU; changes in serum levels are associated with prostate cancer
Catalog No. 50-291-1605 Supplier Cayman Chemical Supplier No. 36181100 MG
A dipeptide with metabolism-altering activity; prevents increases in lipid accumulation in oleic acid-stimulated AML12 hepatocytes at 30 µM; increases CaSR levels and secretion of CCK and GLP-1 in STC-1 enteroendocrine cells at 2.5-10 mM; changes in urine levels are associated with hypertension and PKU; changes in serum levels are associated with prostate cancer