Learn More
Medchemexpress LLC Glu-urea-Glu-NHS ester | 1228927-36-3 | 99.5% | 585.64 | 100 MG
SDP
Catalog No. 5000360778
Encompass_Preferred
Glu-urea-Glu-NHS ester is an activated N-hydroxysuccinamide (NHS) ester of Glu-urea-Glu. It serves as a versatile intermediate for various syntheses.
  • Suitable for the synthesis of small-molecule prostate-specific membrane antigen (PSMA) inhibitors.
  • Can be used in the development of imaging agents, including PSMAi-PEG conjugates.
  • Key component for creating targeted drug delivery vehicles.
  • Functions as an intermediate for synthesizing small-molecule PSMAi-PEG ligands.
  • Enables conjugation to fluorescent nanoparticles for targeted optical imaging of cancer and drug delivery.
Catalog No. 50-003-60778 Supplier Medchemexpress LLC Supplier No. HY147287100MG
Glu-urea-Glu-NHS ester is an activated N-hydroxysuccinamide (NHS) ester of Glu-urea-Glu. It serves as a versatile intermediate for various syntheses.
  • Suitable for the synthesis of small-molecule prostate-specific membrane antigen (PSMA) inhibitors.
  • Can be used in the development of imaging agents, including PSMAi-PEG conjugates.
  • Key component for creating targeted drug delivery vehicles.
  • Functions as an intermediate for synthesizing small-molecule PSMAi-PEG ligands.
  • Enables conjugation to fluorescent nanoparticles for targeted optical imaging of cancer and drug delivery.