Learn More
Medchemexpress LLC Glu-urea-Glu-NHS ester | 1228927-36-3 | 99.5% | 585.64 | 5 MG
SDP
Catalog No. 5000359964
Encompass_Preferred
Glu-urea-Glu-NHS ester is an activated N-hydroxysuccinamide (NHS) ester of Glu-urea-Glu. It is a key intermediate for synthesizing small-molecule prostate-specific membrane antigen (PSMA) inhibitors and imaging agents, as well as for targeted drug delivery vehicles.
  • Utilized in the synthesis of small-molecule PSMA inhibitors.
  • Can be used for imaging agents, such as PSMAi-PEG conjugates.
  • Applicable for targeted drug delivery vehicles.
  • Key intermediate for synthesizing small-molecule PSMAi-PEG ligands.
  • Ligands can be conjugated to fluorescent nanoparticles for targeted optical imaging of cancer and drug delivery.
Catalog No. 50-003-59964 Supplier Medchemexpress LLC Supplier No. HY1472875MG
Glu-urea-Glu-NHS ester is an activated N-hydroxysuccinamide (NHS) ester of Glu-urea-Glu. It is a key intermediate for synthesizing small-molecule prostate-specific membrane antigen (PSMA) inhibitors and imaging agents, as well as for targeted drug delivery vehicles.
  • Utilized in the synthesis of small-molecule PSMA inhibitors.
  • Can be used for imaging agents, such as PSMAi-PEG conjugates.
  • Applicable for targeted drug delivery vehicles.
  • Key intermediate for synthesizing small-molecule PSMAi-PEG ligands.
  • Ligands can be conjugated to fluorescent nanoparticles for targeted optical imaging of cancer and drug delivery.