Learn More
Medchemexpress LLC Glu-urea-Glu-NHS ester | 1228927-36-3 | 99.5% | 585.64 | 50 MG
SDP
Catalog No. 5000359963
Encompass_Preferred
Glu-urea-Glu-NHS ester is an activated N-hydroxysuccinamide (NHS) ester of Glu-urea-Glu. It is primarily used for the synthesis of small-molecule prostate-specific membrane antigen (PSMA) inhibitors, imaging agents, and targeted drug delivery vehicles. In vitro, it can serve as a key intermediate for synthesizing small-molecule PSMAi-PEG ligands, which can be conjugated to fluorescent nanoparticles for targeted optical imaging of cancer and drug delivery.
  • Used for synthesizing small-molecule prostate-specific membrane antigen (PSMA) inhibitors.
  • Can be a key intermediate for synthesizing small-molecule PSMAi-PEG ligands.
  • PSMAi-PEG ligands can be conjugated to fluorescent nanoparticles for targeted optical imaging of cancer and drug delivery.
Catalog No. 50-003-59963 Supplier Medchemexpress LLC Supplier No. HY14728750MG
Glu-urea-Glu-NHS ester is an activated N-hydroxysuccinamide (NHS) ester of Glu-urea-Glu. It is primarily used for the synthesis of small-molecule prostate-specific membrane antigen (PSMA) inhibitors, imaging agents, and targeted drug delivery vehicles. In vitro, it can serve as a key intermediate for synthesizing small-molecule PSMAi-PEG ligands, which can be conjugated to fluorescent nanoparticles for targeted optical imaging of cancer and drug delivery.
  • Used for synthesizing small-molecule prostate-specific membrane antigen (PSMA) inhibitors.
  • Can be a key intermediate for synthesizing small-molecule PSMAi-PEG ligands.
  • PSMAi-PEG ligands can be conjugated to fluorescent nanoparticles for targeted optical imaging of cancer and drug delivery.