Learn More
Bachem (Glu1)-Fibrinopeptide B (human)
SDP
Catalog No. 50259672
Encompass_Preferred
(Glu1)-Fibrinopeptide B (human), H-Glu-Gly-Val-Asn-Asp-Asn-Glu-Glu-Gly-Phe-Phe-Ser-Ala-Arg-OH, 103213-49-6, C66H95N19O26, Mr 1570.59, 1mg.
Catalog No. 50-259-672 Supplier Bachem Supplier No. 4016660.0001
(Glu1)-Fibrinopeptide B (human), H-Glu-Gly-Val-Asn-Asp-Asn-Glu-Glu-Gly-Phe-Phe-Ser-Ala-Arg-OH, 103213-49-6, C66H95N19O26, Mr 1570.59, 1mg.