Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters [Glu1]-Fibrinopeptide B Standard, Mass Spectrometry, 0.1 mg, Solid

Catalog No. 50793036 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Mass Spectrometry standard is designed for system assessment and benchmarking to ensure optimum performance. It contains 0.1 mg of [Glu1]-Fibrinopeptide B in a 4 mL amber screw-top vial and includes a Certificate of Analysis. Fibrinopeptide B (FPB) is composed of 14 amino acids.

  • Designed for system assessment and benchmarking.
  • Contains 0.1 mg of [Glu1]-Fibrinopeptide B (solid formulation).
  • Includes a certificate of analysis.
  • Store at -20°C; 12-month shelf life (1 week after reconstitution).
  • Supports LC-MS analysis to prevent cross-contamination and assess instrument performance.
  • Used for two-point calibration in 1D and 2D gel electrophoresis and protein identification.
  • Corrects mass drift in MS and MS/MS data.

Catalog No. 50-793-036 Supplier Waters Corp Supplier No. 700004729
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This Mass Spectrometry standard is designed for system assessment and benchmarking to ensure optimum performance. It contains 0.1 mg of [Glu1]-Fibrinopeptide B in a 4 mL amber screw-top vial and includes a Certificate of Analysis. Fibrinopeptide B (FPB) is composed of 14 amino acids.

  • Designed for system assessment and benchmarking.
  • Contains 0.1 mg of [Glu1]-Fibrinopeptide B (solid formulation).
  • Includes a certificate of analysis.
  • Store at -20°C; 12-month shelf life (1 week after reconstitution).
  • Supports LC-MS analysis to prevent cross-contamination and assess instrument performance.
  • Used for two-point calibration in 1D and 2D gel electrophoresis and protein identification.
  • Corrects mass drift in MS and MS/MS data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.