Learn More
Medchemexpress LLC (Glu17,21,24)-Osteocalcin (1-49) (human) | 1927927-11-4 | 98.3% | 5797.40 | 100 UG
SDP
Catalog No. 5000396474
Encompass_Preferred
(Glu17,21,24)-Osteocalcin (1-49) (human) is a bone-specific protein primarily involved in critical biological processes such as bone formation and glucose metabolism. It is provided as a solid, white to off-white substance for research applications.
  • Bone-specific protein
  • Involved in bone formation
  • Involved in glucose metabolism
  • High purity of 98.3%
  • Features a disulfide bridge between Cys23 and Cys29
  • For research use only
Catalog No. 50-003-96474 Supplier Medchemexpress LLC Supplier No. HYP4588100UG
Add to Cart
Add to Cart
(Glu17,21,24)-Osteocalcin (1-49) (human) is a bone-specific protein primarily involved in critical biological processes such as bone formation and glucose metabolism. It is provided as a solid, white to off-white substance for research applications.
  • Bone-specific protein
  • Involved in bone formation
  • Involved in glucose metabolism
  • High purity of 98.3%
  • Features a disulfide bridge between Cys23 and Cys29
  • For research use only