Learn More
Bachem (Glu2)-TRH
SDP
Supplier:  Bachem 4026102.0050
Encompass_Preferred
(Glu2)-TRH, Pyr-Glu-Pro-NH2, 85541-78-2, C15H22N4O6, Mr 354.36, 50mg. Fertilization-Promoting Peptide, FPP, pE-E-P-NH2. Fertilization-promoting peptide (FPP), a TRH analog, was previously purified and characterized from both rabbit prostate and human semen. Nanomolar concentrations of this peptide have been shown to enhance capacitation of mouse epididymal spermatozoa with a concomitant increase in fertilizing ability. It was also detected in the thyroid gland. According to a study of Nguyen et al. Pyr-Glu-Pro-NH2 opposes the cholinergic effect of TRH in the mammalian CNS: Co-perfusion with an equivalent of TRH in the rat hippocampus resulted in a significant attenuation of TRH-induced acetylcholine release.
Catalog No. 50-382-569