Learn More
American Radiolabeled Chemicals Inc GLUCAGON 125I 50UC
SDP
Catalog No. 50901016
Encompass
Glucagon 125I receptor grade 50uCi in buffered solution, 0.08 mCi/ml, S.A. 2200 Ci/mmol, M.W. 3608
Catalog No. 50-901-016 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 013950UC
Add to Cart
Add to Cart
Request a Quote
Glucagon 125I receptor grade 50uCi in buffered solution, 0.08 mCi/ml, S.A. 2200 Ci/mmol, M.W. 3608