Learn More
Bachem Glucagon (19-29) (human, rat, porcine) Trifluoroacetate
SDP
Supplier:  Bachem 4030650.0005
Encompass_Preferred
Glucagon (19-29) (human, rat, porcine) Trifluoroacetate, H-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-OH, 64790-15-4, C61H89N15O18S, Mr 1352.53, 5mg. Miniglucagon (human, rat, porcine). Miniglucagon inhibited both the Ca2+ activated and Mg2+ dependent ATPase activity and Ca2+ transport in liver plasma membranes with an efficiency 1000-fold higher than that of glucagon. It is likely to be the active peptide involved in the inhibition of the liver Ca2+ pump. Miniglucagon, the ultimate processing product of proglucagon, is a regulator of the pancreatic islet physiology.
Catalog No. 50-382-618