Learn More
Medchemexpress LLC HY-P0082 50mg , Glucagon Porcine glucagon CAS:16941-32-5 Purity:98%
SDP
Supplier:  Medchemexpress LLC HYP008250MG
Encompass_Preferred
Medchemexpress, HY-P0082 50mg Glucagon Porcine glucagon CAS:16941-32-5 Formula:C153H225N43O49S Purity:98% Glucagon is a peptide hormone, exhibits therapeutic potential for metabolic disease. Sequence: His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3791