Learn More
Mercodia Inc Glucagon Control, Low Medium High, For Glucagon ELISA
SDP
Catalog No. NC1062190
Encompass_Preferred
Control set containing synthetic glucagon. Three-level control: Low, Medium, High. ADVANTAGESEasy to transport ¿ Lyophilized bufferEasy to use ¿ Reconstituted with redistilled water
Catalog No. NC1062190 Supplier Mercodia Inc Supplier No. 10128601
Control set containing synthetic glucagon. Three-level control: Low, Medium, High. ADVANTAGESEasy to transport ¿ Lyophilized bufferEasy to use ¿ Reconstituted with redistilled water