Learn More
Ansh Labs Ansh Labs Glucagon ELISA kit, 96-well colorimetric assay (FDA registered). Detection in plasma in human, goat, rabbit, bovine, equine, ovine, porcine, canine, mouse, squirrel monkey & vervet monkey. Range of 7-314 pg/mL & sensitivity of 2.4 pg/mL.
SDP
Supplier:  Ansh Labs AL157
Encompass
The Glucagon ELISA kit measures Glucagon with no detectable cross-reactivity to Glicentin, Oxyntomodulin, GLP-1, GLP-2, or GRPP. The kit has species reactivity to human, goat, rabbit, bovine, equine, ovine, porcine, canine, mouse, squirrel monkey and vervet monkey samples. The kit offers an analytical range of 7-314 pg/mL and a sensitivity of 2.4 pg/mL, using 50 uL of plasma. The kit is an HRP-based ELISA with colorimetric detection by dual wavelength absorbance at 450 nm and 630 nm as reference filter and has an assay time of 2.5 hours.Intended for In vitro Diagnostic Use.
Catalog No. 50-194-3737