Learn More
Crystal Chem Inc Glucagon ELISA Kit, 96-wells, Uses 10uL of sample
SDP
Supplier:  Crystal Chem Inc 81520
Encompass
The Glucagon ELISA is used to quantify levels of human glucagon in plasma, serum, and cell culture using only a 10 µL sample volume. Highlights: Kits use a sample volume of only 10 µL. High sensitivity of 1.1 pg/mL. Dynamic Range of 7.8 - 500 pg/mL. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7905