Learn More
Biomatik Corporation Glucagon (GCG) ELISA Kit
Supplier:  Biomatik Corporation EKU12387
Encompass_Preferred
Glucagon (GCG) ELISA Kit for detecting GCG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 18.52-1,500pg/ml. Sensitivity: 6.94pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: GC; GLP1; GLP2; GRPP; Glicentin-Related Polypeptide; Glucagen; Oxyntomodulin; Incretin hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5004