Learn More
Medchemexpress LLC HY-P1145A 1mg Medchemexpress, Glucagon-like peptide 1 (1-37), human (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1145A1MG
Encompass_Preferred
Medchemexpress, HY-P1145A 1mg Glucagon-like peptide 1 (1-37), human (TFA) CAS: Glucagon-like peptide 1 (1-37), human (TFA) is a highly potent agonist of the GLP-1 receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0526