Learn More
Bachem GLP-1 (1-37) (human, bovine, guinea pig, mouse, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4037542.1000
Encompass_Preferred
GLP-1 (1-37) (human, bovine, guinea pig, mouse, rat) Trifluoroacetate, H-His-Asp-Glu-Phe-Glu-Arg-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-Gly-OH, 87805-34-3, C186H275N51O59, Mr 4169.54, 1mg. Proglucagon (72-108) (human, bovine, guinea pig, mouse, rat), Glucagon-Like Peptide 1 (1-37) (human, bovine, guinea pig, mouse, rat), Preproglucagon (92-128) (human, bovine, guinea pig, mouse, rat). Glucagon-Like Peptide 1 (GLP-1) is synthesized by posttranslational processing of proglucagon in the intestine and pancreas and plays an important role in metabolic homeostasis. Among the different molecular forms, such as GLP-1 (7-36) amide and GLP-1 (7-37), the function of GLP-1 (1-37) has been unclear.
Catalog No. 50-194-6893