Learn More
Medchemexpress LLC Glucagon-like peptide-1 (GLP-1)(7-36), amide | 107444-51-9 | 99.9% | 3297.68 | 1 MG
SDP
Catalog No. 5000426210
Encompass_Preferred
Glucagon-like peptide-1 (GLP-1)(7-36), amide is a physiological incretin hormone that stimulates insulin secretion. This peptide has a molecular weight of 3297.68 and appears as a white to off-white solid.
  • Stimulates insulin secretion.
  • Potent insulin-releasing peptide.
  • Product of tissue-specific post-translational processing.
  • Released postprandially from intestinal endocrine L cells.
  • Normalizes fasting plasma glucose concentrations in Type 2 diabetic subjects (exogenous).
  • Soluble in H2O at 100 mg/mL (30.32 mM; requires ultrasonic).
  • For research use only.
Catalog No. 50-004-26210 Supplier Medchemexpress LLC Supplier No. HYP0054A1MG
Glucagon-like peptide-1 (GLP-1)(7-36), amide is a physiological incretin hormone that stimulates insulin secretion. This peptide has a molecular weight of 3297.68 and appears as a white to off-white solid.
  • Stimulates insulin secretion.
  • Potent insulin-releasing peptide.
  • Product of tissue-specific post-translational processing.
  • Released postprandially from intestinal endocrine L cells.
  • Normalizes fasting plasma glucose concentrations in Type 2 diabetic subjects (exogenous).
  • Soluble in H2O at 100 mg/mL (30.32 mM; requires ultrasonic).
  • For research use only.