Learn More
Medchemexpress LLC Glucagon-like peptide-1 (GLP-1)(7-36), amide acetate | 1119517-19-9 | 99.0% | 3297.64 (free base) | 1 MG
SDP
Catalog No. 5000426682
Encompass_Preferred
GLP-1(7-36), amide acetate is a major intestinal hormone that stimulates glucose-induced insulin secretion from β cells. It is for research use only.
  • Stimulates glucose-induced insulin secretion from β cells.
  • Active GLP-1 secretion is stimulated by fatty acids such as palmitic, oleic, linoleic, or linolenic acid.
  • Increases active GLP-1 concentrations in vitro.
  • Increases active GLP-1(7-36) amide levels in portal blood in vivo.
  • Co-administration of glucose and tea oil additively increases peak GLP-1 levels.
Catalog No. 50-004-26682 Supplier Medchemexpress LLC Supplier No. HYP00541MG
GLP-1(7-36), amide acetate is a major intestinal hormone that stimulates glucose-induced insulin secretion from β cells. It is for research use only.
  • Stimulates glucose-induced insulin secretion from β cells.
  • Active GLP-1 secretion is stimulated by fatty acids such as palmitic, oleic, linoleic, or linolenic acid.
  • Increases active GLP-1 concentrations in vitro.
  • Increases active GLP-1(7-36) amide levels in portal blood in vivo.
  • Co-administration of glucose and tea oil additively increases peak GLP-1 levels.