Learn More
Genscript Corporation Glucagon-Like Peptide (GLP) II, human
Supplier:  Genscript Corporation RP107690.5
Encompass
Glucagon-like peptide 2 (GLP-2) is a recently identified intestinal epithelium-specific growth factor that has been shown to reduce the severity of inflammatory disorders of the intestine in rodent models. Currently Glucagon-Like Peptide 2 is used as a potential therapeutic agent for the human subjects with a broad variety of intestinal diseases characterized by intestinal damage and insufficiency.
Catalog No. 50-274-944