Learn More
Echelon Biosciences Research Labs Glucagon-like Peptide I human / Preproglucagon (72-107), amide / GLP 1, human - 0.5 mg, CAS#: 99658-04-5
SDP
Supplier:  Echelon Biosciences Research Labs 471270.5MG
Encompass_Preferred
Glucagon-like Peptide I human / Preproglucagon (72-107), amide / GLP 1, human - 0.5mg , His-Asp-Glu-Phe-Glu-Arg-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys-Glu-Phe-Ile-Ala-Trp-Leu-Val-Lys-Gly-Arg-NH2 Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8486