Learn More
Enzo Life Sciences Glucagon monoclonal antibody (09) (1 mg)
SDP
Supplier:  Enzo Life Sciences BPDABS061091
Encompass_Preferred
Clone: 09. Isotype: IgG1κ. Immunogen: Synthetic C-terminal fragment corresponding to human glucagon coupled to carrier. Species reactivity: Human, Rat Porcine. Applications: ELISA. Purity: Protein A-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-201-2438