Learn More
Cayman Chemical Glucagon RECPTR EXrAClul 100ug
SDP
Catalog No. 502943399
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged GCGR extracellular domain expressed in HEK293 cells • Amino acids: 26-136 • MW: 14.5 kDa
Catalog No. 50-294-3399 Supplier Cayman Chemical Supplier No. 42231100 UG
Source: Active recombinant human C-terminal His-tagged GCGR extracellular domain expressed in HEK293 cells • Amino acids: 26-136 • MW: 14.5 kDa